Eyewash with Drench Hose, Speakman®
Supplier: Speakman
SE-927
VF-0927-SEEA 1712 USD
VF-0927-SE
Eyewash with Drench Hose, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
The eyewash features a stay-open valve and twin aerated sprays with flip-top dust caps and automatic flow control.
  • Eyewash is supplied with 1.8 m (6') of reinforced hose
Designed for countertop mounting, the unit can also be used as a handheld drench hose.
Certifications: Meets ANSI Z358.1 standard.