KEMRESIN® and KEMSHIELD® Polishes
Supplier: Kewaunee
0065-A1
VF-0065-A1EA 80.86 USD
VF-0065-A1 VF-0065-A2
KEMRESIN® and KEMSHIELD® Polishes
Cleaning Agents Furniture Polish
Apply polishes to work surfaces and shelving following detergent cleaning.
Helps keep surfaces clean, and hides minor scratches and blemishes.