Florida TRPH Standard (17 Components), Ultra Scientific
Supplier: AGILENT TECHNOLOGIES
SFL-601-1
ULSFL-601-1EA 117.17 USD
ULSFL-601-1
Florida TRPH Standard (17 Components), Ultra Scientific
Standards Organic Standards
This contains n-octane (C₈), n-decane (C₁₀), n-dodecane (C₁₂), n-tetradecane (C₁₄), n-hexadecane (C₁₆), n-octadecane (C₁₈), n-eicosane (C₂₀), n-docosane (C₂₂), n-tetracosane (C₂₄), n-hexacosane (C₂₆), n-octacosane (C₂₈), n-triacontane (C₃₀), n-dotriacontane (C₃₂), n-tetratriacontane (C₃₄), n-hexatriacontane (C₃₆), n-octatriacontane (C₃₈) and n-tetracontane (C₄₀) at 500 µg/ml in hexane.