IC Anions Standard, Mix 2, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES
ICC-210
ULICC-210EA 520.19 USD
ULICC-210
IC Anions Standard, Mix 2, Agilent Technologies
Standards Anionic Standards Multi-Element Anioinic Standards
Multi component mixture.
  • Solvent: Water (low TOC, 50 ppb)
  • Agency method: EPA 300.1, EPA 300 and EPA 9056A