CleanTips® Micro Alpha® ESD Swab, Texwipe®
Supplier: Texwipe
TX758E
TWTX758ECS 650.94 USD
TWTX758E
CleanTips® Micro Alpha® ESD Swab, Texwipe®
Swabs Swabs for Cleaning
The Micro Alpha® ESD Swab is an excellent dissipative swab for cleaning very small grooved, slotted, and recessed areas
Ideal for use with many cleaning solvents and solutions. A compact, electrically dissipative handle and flexible internal head paddle provide support and control. The laundered Alpha® polyester head is extremely clean. Swab head features ultraclean, cleanroom laundered, double-layer, knitted Alpha® polyester with flexible internal paddle. Swab handle is made of low-contamination Transplex®, a unique plastic alloy that safely dissipates electric charges without contaminating additives. Swab handle bears the ESD-safe imprint.
Ordering information: Available in bag or case quantities.