CleanTips® CleanFoam® Medium Compressed ESD Swab
Supplier: Texwipe
TX752E
TWTX752EBG 77.29 USD
TWTX752E
CleanTips® CleanFoam® Medium Compressed ESD Swab
Swabs Swabs for Cleaning
Texwipe's ESD-Safe swab series features a Stat-Rite® handle, a patented inherently dissipative polymer, to provide clean ESD protection without contributing particulate contaminants to the process or environment. Thermal bond construction eliminates the risk of adhesive contamination.
  • ESD protection with Stat-Rite® handle
  • No blooming carbon particles, metals, or ionic anti-static contaminants
  • Manufactured to exacting and consistent tolerances using high-precision automated processes
  • Packaged in an anti-static bag
  • Absorbs solvents, solutions, and grabs particulates
  • Lot coded for traceability and quality control
Applications are applying and removing lubricants, adhesives, and other solutions in a critical clean environment with ESD concerns; scrubbing recessed areas; removal of excess materials or debris; cleaning intersecting surfaces and joints; cleaning with compatible solutions and solvents; picking up fine powders; appropriate for use with temperatures less than 90 °C/194 °F.
Three designs are available:
• Polyester - Manufactured from laundered Alpha® 100% knit polyester, making the head extremely clean
• Foam - Made from Texwipe's CleanFoam®, cleanroom processed 100 ppi polyurethane foam
• Dual micro-tip pick - made from Stat Rite® polymer only
Applying in industres like aerospace, animal laboratory, biologics, data storage, microelectronics, printing/graphics, medical devices, cleanroom design/build.
Delivery information: Shelf life: 5 years from date of manufacture.