CleanFoam® ESD Swab Mini Compressed, Texwipe®
Supplier: Texwipe
TX750E
TWTX750ECS 763.45 USD
TWTX750E
CleanFoam® ESD Swab Mini Compressed, Texwipe®
Swabs Swabs for Cleaning
The Mini Compressed CleanFoam® ESD Swab is an excellent dissipative swab with a precision, pointed tip that is ideal for cleaning small slotted and grooved areas
These swabs are made of foam.
Its compressed, laundered open-cell CleanFoam® head offers reliable firmness and a pointed tip. A compact, rigid handle and tapered internal core provide good support. The handle is made from a Stat-Rite® polymer alloy using a patented Inherently Dissipative Polymer (IDP) to provide clean, permanent ESD protection without contributing particulate contaminates to the cleanroom. Swab handle bears the ESD-safe imprint.
Ordering information: Available in bag or case quantities.