CleanTips®, Nylon MicroSwab™, Texwipe®
Supplier: Texwipe
TX730
TWTX730CS 552.24 USD
TWTX730
CleanTips®, Nylon MicroSwab™, Texwipe®
Swabs Swabs for Cleaning
These precision nylon swabs are made of 0.9 mm diameter tip constructed of bundled nylon filaments.
  • Crimped construction eliminates adhesive contamination
  • Very low in both non volatile residue and particle generation
  • 100% polypropylene handle
  • Long, thin bundled filaments reach into narrow channels, corners, edges for cleaning particles and removing excess solvent
Designed for precision cleaning of optics and optical fibres, microscopes, reticles, photomasks, and wafers. Also suitable for precision application of solvents and cleaning solutions.
Ordering information: Packaged in a reclosable bag.