CrushTube™ Swab, Texwipe®
Supplier: Texwipe
TX726
TWTX726CS 996.59 USD
TWTX726
CrushTube™ Swab, Texwipe®
Swabs Swabs for Cleaning
Ideal for cleaning ionizer tips
Point-of-use IPA-saturated polyester swab has a polyester brush tip with a breakable glass ampule containing 91% isopropyl alcohol (IPA). When ampule is crushed using protective plastic sleeve, the brush tip becomes saturated and is ready for use. Silicone-free.
Ordering information: Packaged 50 swabs per bag, 10 boxes per case for a total of 500 swabs per case. Bag-Within-A-Bag® packaging.