CleanTips® Flexible Medium Head Foam Swab, Texwipe®
Supplier: Texwipe
TX709A
TWTX709ACS 574.02 USD
TWTX709A
CleanTips® Flexible Medium Head Foam Swab, Texwipe®
Swabs Swabs for Cleaning
Flexible, general-purpose swab for cleaning surfaces and recessed areas
Swab features a medium head on a flexible nylon paddle. Made from closed-cell polyurethane foam with 100 pore-per-inch porosity, this swab possesses excellent durability and cushioning. Thermal and epoxy head seal. An internal paddle provides flexible support. High-grade polyurethane foam head will not crack or chip. Low particle and fiber generation, and good chemical resistance.
The round, light green polypropylene handle enables good control and ease of use.
Can be used in a wide variety of industrialMore Product Information