Sterile TexTra™ Wipers, Texwipe®
Supplier: Texwipe
TX3220
TWTX3220CS 674.22 USD
TWTX3220
Sterile TexTra™ Wipers, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Sterile polyester wipers are effective for cleaning equipment and environmental surfaces in critical aseptic cleanrooms without compromising the sterile environment.
  • Single-ply
  • Double-knit
  • Cleanroom laundered
Suitable for the final rinsing and residue removal step of most critical cleaning protocols. Wipers are made of 100% continuous-filament polyester, have sealed borders.
Certifications: Lot-specific Certificates of Processing and Compliance are included. Expiration dates are marked on each bag. For use in ISO Class 5 (FED-STF-209E Class 100/M3.5) cleanroom environments. Gamma irradiated to a Sterility Assurance Level of 10⁻⁶.
Ordering information: Double-bagged in solvent safe bags that can be wiped down with alcohol before introduction of inner bag to sterile environments.
Packaging: Packaged 100 wipers/bag (5 packs of 20 wipers); 5 bags per case (500 wipers/case).