SterileWipe™ Low Particle Wiper, Texwipe®
Supplier: Texwipe
TX3211
TWTX3211CS 651.97 USD
TWTX3211
SterileWipe™ Low Particle Wiper, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
SterileWipe™ Low Particle Wiper is a 100% continuous-filament polyester wiper that is low in particle generation and extractable levels and provides excellent cleaning performance in critical sterile environments
Ideal for use when bacteriological contamination, particulates, and extractables are critical factors, and ideal for environments where low particulate levels are most critical. Suitable for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) environments.
Each package is gamma irradiated to 10–6 Sterility Assurance Level.
Designed for cleaning sterile environments and dispensing solutions.
Ordering information: Packaged 10 wipers per pack, 10 packs per bag in Solvent Safe packaging.