Vectra® Honeycomb® 10 Cleanroom Wiper, Texwipe®
Supplier: Texwipe
TX1060
TWTX1060CS 1236.54 USD
TWTX1060
Vectra® Honeycomb® 10 Cleanroom Wiper, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Vectra® Honeycomb® 10 dry cleanroom wiper is made from 100% polyester with a sealed border, processed on Texwipe’s automated cleaning system.
  • Vectra sealed border technology provides low levels of ions, NVRs (non volatile residues), particles and fibers for use in critical cleaning applications and environments
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Appropriate for use with temperatures less than 400°F (205°C)
  • Autoclave safe
  • Individually lot coded for ease of traceability and quality control
The Vectra® Honeycomb® 10 is designed to combine the benefits of two wipers in one. One side contains honeycomb pockets for capturing and retaining particles including debris. The other side is smooth. Together, they form a high sorption wiper ideal for spill control, cleaning, and solution application.
Meets USP <797> and USP <800> wiper requirements.
Cleanroom Environment: ISO Class 3 - 8, Class 1 -100,000, EU Grade A - D
Packaging: Inner Bags: 100 wipers/bag (2 inner bags of 50 wipers). Outer Bags: 10 bags/case. Wipers Total: 1,000 wipers total/case.