TexWrite® Clipboards, Texwipe®
Supplier: Texwipe
TX5835
TW-TX5835CS 474.31 USD
TW-TX5835
TexWrite® Clipboards, Texwipe®
Clipboards
Ideal for portable writing needs in critical environments, this unique and durable clipboard is constructed of rigid polypropylene with a durable plastic clip mechanism for longlasting use
  • Rigid polypropylene
  • Chemical resistant
  • Cleanroom packaged
  • Securely holds 1 – 50 sheets of cleanroom paper
  • Rounded corners, no sharp edges to damage gloves or garments
Can be used with 8.5”x11” and A4 paper.