SureSTART™ Caps for 2 ml Polypropylene Snap Top Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
6ERC11PE
TS6ERC11PEPK 13.33 USD
TS6ERC11PE
SureSTART™ Caps for 2 ml Polypropylene Snap Top Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Closures Chromatography Vial Closures
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml polypropylene snap top microvials for everyday chromatography analysis of <2 ml samples. They provide an affordable choice and can be used with all HPLC/UHPLC instrument types.
These clear plastic 2 ml snap top vials are specification certified to ensure autosampler compatible physical dimensions. Seal these snap top vials with 11 mm snap caps and septa.