Acetonitrile for LC-MS

Supplier: Thermo Scientific Chemicals
Danger

61514-0025
TS61514-0025EA 672 USD
TS61514-0025
Acetonitrile for LC-MS
Acetonitrile
Formula: H₃CCN
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR