Amphotericin B

Supplier: Thermo Scientific Chemicals

61412-2500
TS61412-2500EA 169 USD
TS61412-2500
Amphotericin B
Amphotericin B
Formula: C₄₇H₇₃NO₁₇
Storage Temperature: Ambient
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now

Specification Test Results

Appearance Yellow Powder
Loss on drying ≤5.0 % ()
Additional info amphotericin A: < 15%
Identification passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR