Calcium stearate USP

Supplier: Thermo Scientific Chemicals

Synonyms: Stearic acid calcium salt

61243-0030
TS61243-0030EA 177 USD
TS61243-0030
Calcium stearate USP
Calcium stearate
Formula: C₃₆H₇₀CaO₄
MDL Number: MFCD00036390
CAS Number: 1592-23-0

Order Now

Specification Test Results

Appearance White Granular powder
Identification passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR