Sand, quartz, white (Sand)

Supplier: Thermo Scientific Chemicals

61235-0050
TS61235-0050EA 140 USD
TS61235-0050
Sand, quartz, white (Sand)
Sand, quartz
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7

Order Now

Specification Test Results

Appearance Light grey to tan Crystals
Identification passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR