Barium chloride, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

AC61228-1000
TS61228-1000EA 206 USD
TS61228-1000
Barium chloride, anhydrous
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: ∼963 °C
Density: 2.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998

Order Now

Specification Test Results

Appearance White Granules
Assay ≥97 % (BaCl2)
Identification passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR