Streptomycin sulfate

Supplier: Thermo Scientific Chemicals
Warning

61224-0500
TS61224-0500EA 61 USD
TS61224-0500
Streptomycin sulfate
Streptomycin sulphate
Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457.4 g/mol
Melting Pt: 250
Storage Temperature: Refrigerator
MDL Number: MFCD00037023
CAS Number: 3810-74-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,08983

Order Now

Specification Test Results

Appearance White to light yellow Powder
Loss on drying ≤5 %
Identification passes test
pH 4.5 to 7.0 (20 % solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR