Eriochrome Black T solution

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

61222-5000
TS61222-5000EA 77 USD
TS61222-5000
Eriochrome Black T solution
Eriochrome Black T
Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear blue Solution
Identification passes test
Sensitivity passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR