Ammonium iron (III) citrate 16.5 - 18.5% (Fe)

Supplier: Thermo Scientific Chemicals
Warning

AC61221-5000
TS61221-5000EA 68 USD
TS61221-5000
Ammonium iron (III) citrate 16.5 - 18.5% (Fe)
Ammonium iron (III) citrate
Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6

Order Now

Specification Test Results

Color Brown to Brown-Yellow
Physical Form Powder
Percent Purity 16.5 to 18.5% (Fe)
Identification Pass Test
Infrared Spectrum Authentic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR