Zinc stearate

Supplier: Thermo Scientific Chemicals

AC61151-5000
TS61151-5000EA 30 USD
TS61151-5000
Zinc stearate
Zinc stearate
Formula: C₃₆H₇₀O₄Zn
Melting Pt: 118…128 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Assay 12.5 to 14.0 % (ZnO)
Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR