Dextran sulfate sodium salt

Supplier: Thermo Scientific Chemicals

Synonyms: Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran

61136-1000
TS61136-1000EA 271 USD
TS61136-1000
Dextran sulfate sodium salt
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Loss on drying ≤7.5 % ()
Additional info sulfur: 17 - 20 %
Identification passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR