Kanamycin sulfate

Supplier: Thermo Scientific Chemicals
Danger

61129-0050
TS61129-0050EA 138 USD
TS61129-0050
Kanamycin sulfate
Kanamycin sulphate
Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Appearance White Powder
Additional info chromatographic purity: passes test
Identification passes test
pH 6.5 to 8.5 (1 % solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR