Silicic acid hydrate, powder

Supplier: Thermo Scientific Chemicals

AC61123-5000
TS61123-5000EA 379 USD
TS61123-5000
Silicic acid hydrate, powder
Silicic acid hydrate

Appearance: White Powder

Formula: SiO₂·xH₂O
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS Number: 1343-98-2
REACH: 01-2119379499-16

Order Now

Specification Test Results

Appearance White Powder
Identification Passes test
Non-volatile matter ≤0.40 % (in HF)
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.020 %
Iron (Fe) ≤0.01 %
Heavy metals (as Pb) ≤0.010 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR