Electrode Storage Solution, Thermo Scientific
Supplier: Thermo Scientific Chemicals
61121-0010
TS61121-0010EA
100
USD
TS61121-0010
Electrode Storage Solution, Thermo Scientific
Electrolytes
Electrode Storage Solutions
CAS No.:
MDL No.: MFCD00211908
Density: 1
Boiling Point: 100 °C (760 mm Hg)
Appearance: Clear colorless Liquid
Storage Temperature: Ambient
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...