Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)

Supplier: Thermo Scientific Chemicals

61105-0040
TS61105-0040EA 264 USD
TS61105-0040
Ethanol denatured for HPLC (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured
Formula: H₃CCH₂OH
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Refractive index 1.3580 to 1.3610 (25 °C, 589 nm)
Assay 89 to 91 %
Water ≤0.10 %
Additional info LC use test: passes test
Solubility (in water) Passes test
UV at 254 nm A: ≤0.04
Color scale ≤5 APHA
Identification passes test
Residue after evaporation ≤5 ppm
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
Methanol 4.0 to 5.0 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR