Tetrahydrofuran, extra dry, anhydrous ≥99.9%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

61092-1000
TS61092-1000EA 85 USD
TS61092-1000
Tetrahydrofuran, extra dry, anhydrous ≥99.9%, AcroSeal™
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless Liquid
Assay ≥99.9 %
Water ≤50 ppm
Identification passes test
Peroxides ≤0.015 % (as THF peroxide)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR