Tetrahydrofuran stabilized

Supplier: Thermo Scientific Chemicals
Danger

61090-0010
TS61090-0010EA 127 USD
TS61090-0010
Tetrahydrofuran stabilized
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless Liquid
Assay ≥99.9 %
Water ≤50 ppm
Additional info inhibitor (BHT): 75 - 400 ppm
Identification passes test
Peroxides ≤5 ppm (as THF peroxide)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR