Tetrahydrofuran

Supplier: Thermo Scientific Chemicals
Danger

61010-0040
TS61010-0040EA 513 USD
TS61010-0040
Tetrahydrofuran
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Refractive index 1.4040 to 1.4070 (25 °C, 589 nm)
Water ≤0.02 %
Additional info fluorescence background: passes test
GC ≥99.9 %
UV at 254 nm A: ≤0.1
Color scale ≤10 APHA
Identification passes test
Residue after evaporation ≤5 ppm
Peroxides ≤0.015 % (THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR