Methanol ≥99.9% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

61009-0040
TS61009-0040EA 211 USD
TS61009-0040
Methanol ≥99.9% for HPLC
Methanol
Formula: CH₃OH
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Refractive index 1.3260 to 1.3300 (25 °C, 589 nm)
Water ≤0.1 %
Additional info fluorescence background: passes test
Solubility (in water) Passes test
GC ≥99.9 %
UV at 254 nm A: ≤0.025
Color scale ≤5 APHA
Identification Passes test
Residue after evaporation ≤3 ppm
Gradient analysis Passes test
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
Acetone ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR