1-Butanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

61002-0040
TS61002-0040EA 398 USD
TS61002-0040
1-Butanol
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Refractive index 1.3952 to 1.3992 (25 °C, 589 nm)
Water ≤0.1 %
Additional info fluorescence background: passes test
GC ≥99.4 %
UV at 254 nm A: ≤0.025
Color scale ≤10 APHA
Identification passes test
Residue after evaporation ≤50 ppm
Titrable acid ≤0.0008 meq/g
Aldehyde (per ACS) passes test
Butyl ether ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR