AmberChrom™ chloride form 1X8 200-400 mesh

Supplier: Thermo Scientific Chemicals

472970025 472971000 472975000
TS472970025EA 2150 USD
TS472970025 TS472971000 TS472975000
AmberChrom™ chloride form 1X8 200-400 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
CAS Number: 69011-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR