AmberChrom™ chloride form 1X8 200-400 mesh
Supplier: Thermo Scientific Chemicals
472970025
472971000
472975000
TS472970025EA
2150
USD
TS472970025
TS472971000
TS472975000
AmberChrom™ chloride form 1X8 200-400 mesh
AmberChrom™ chloride form
Formula:
(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x |
CAS Number:
69011-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...