AmberChrom™ chloride form 1X8 100-200 mesh
Supplier: Thermo Scientific Chemicals
472960025
472961000
472965000
TS472960025EA
1361
USD
TS472960025
TS472961000
TS472965000
AmberChrom™ chloride form 1X8 100-200 mesh
AmberChrom™ chloride form
Formula:
(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x |
CAS Number:
69011-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...