AmberChrom™ chloride form 1X8 100-200 mesh

Supplier: Thermo Scientific Chemicals

472960025 472961000 472965000
TS472960025EA 1361 USD
TS472960025 TS472961000 TS472965000
AmberChrom™ chloride form 1X8 100-200 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
CAS Number: 69011-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR