AmberChrom™ chloride form 1X2 50-100 mesh

Supplier: Thermo Scientific Chemicals

472940025 472941000 472945000
TS472940025EA 1966 USD
TS472940025 TS472941000 TS472945000
AmberChrom™ chloride form 1X2 50-100 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
CAS Number: 69011-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR