Filgotinib (GLPG0634) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(5-(4-((1,1-Dioxidothiomorpholino)methyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide, Filgotinib

468181000
TS468181000EA 222 USD
TS468181000
Filgotinib (GLPG0634) 95%
Filgotinib (GLPG0634)

Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.


Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies.

Formula: C₂₁H₂₃N₅O₃S
Storage Temperature: Freezer
MDL Number: MFCD20527867
CAS Number: 1206161-97-8

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR