Pinometostat (EPZ-5676) 95%

Supplier: Thermo Scientific Chemicals
Warning

468161000
TS468161000EA 690 USD
TS468161000
Pinometostat (EPZ-5676) 95%
Pinometostat (EPZ-5676)

Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.


Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies.

Formula: C₃₀H₄₂N₈O₃
Storage Temperature: Freezer
MDL Number: MFCD24849416
CAS Number: 1380288-87-8

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR