NVP-BGJ398 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-[6-[4-(4-ethylpiperazin-1-yl)anilino]pyrimidin-4-yl]-1-methylurea, 3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-(6-((4-(4-ethylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)-1-methylurea, Infigratinib

468101000
TS468101000EA 454 USD
TS468101000
NVP-BGJ398 95%
NVP-BGJ398

Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.


Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies.

Formula: C₂₆H₃₁Cl₂N₇O₃
Storage Temperature: Freezer
MDL Number: MFCD22123241
CAS Number: 872511-34-7

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR