BIX 01294 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine, BIX-01294, N-(1-Benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine

467831000
TS467831000EA 260 USD
TS467831000
BIX 01294 95%
BIX 01294

Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.


Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies.

Formula: C₂₈H₃₈N₆O₂
Storage Temperature: Freezer
MDL Number: MFCD14560563
CAS Number: 935693-62-2

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR