Pirfenidone
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone
46472-0010
46472-2500
TS46472-0010EA
354
USD
TS46472-0010
TS46472-2500
Pirfenidone
Pirfenidone
Formula:
C₁₂H₁₁NO MW: 185.23 g/mol Melting Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00866047 CAS Number: 53179-13-8 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 107 to 111 °C |
Loss on drying | ≤0.5 % (1 g, 80 °C, 4 hr, 75 kPa) |
Solubility | (5% in CH2Cl2) Clear colorless to yellow |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...