Ammonium chloride for molecular biology

Supplier: Thermo Scientific Chemicals
Warning

46459-5000
TS46459-5000EA 128 USD
TS46459-5000
Ammonium chloride for molecular biology
Ammonium chloride

  • Bioactive

Formula: NH₄Cl
Melting Pt: 340 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9

Order Now

Specification Test Results

Appearance White Crystalline powder, crystals and/or chunks
Solubility (10 % in water) Clear colorless solution
Titration Argentimetric ≥99.4 %
pH 4.5 to 5.5
Dnase,Rnase,Protease activity none detected
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR