Chloroform-[D₁] 99.8 atom % D + 0.1 v/v% TMS stabilized for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with silver

464430250
TS464430250EA 35 USD
TS464430250
Chloroform-[D₁] 99.8 atom % D + 0.1 v/v% TMS stabilized for NMR spectroscopy
Chloroform-[D1]

  • Bioactive

Formula: CDCl₃
Boiling Pt: 60 °C (760 mmHg)
Melting Pt: –64 °C
Storage Temperature: Ambient
MDL Number: MFCD00000827
CAS Number: 865-49-6
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 %
D-Enrichment ≥99.75 %
TMS 0.09 to 0.11 v/v %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR