Nitromethane-[D₃] 99.5 atom % D NMR

Supplier: Thermo Scientific Chemicals

Synonyms: Nitromethane-[²H₃]

464230050
TS464230050EA 96 USD
TS464230050
Nitromethane-[D₃] 99.5 atom % D NMR
Nitromethane-[D3]
Formula: D₃CNO₂
MW: 64.02 g/mol
Boiling Pt: 100
Melting Pt: -26
MDL Number: MFCD00044214
CAS Number: 13031-32-8
UN: 1261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR