Acetonitrile-[D₃] 100 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

464170075
TS464170075EA 193 USD
TS464170075
Acetonitrile-[D₃] 100 atom % D for NMR spectroscopy
Acetonitrile-[D3]
Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7
Melting Pt: -42
Flash Pt: 2
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR