Acetic acid-[D₄] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

Synonyms: Ethanoic acid-D4, Acetic-D3 acid-D, Acetic-d3 acid-d, Acetic acid-[²H₄], Tetradeuteroacetic acid, Acetic acid-d₄

464090075
TS464090075EA 81 USD
TS464090075
Acetic acid-[D₄] 99.5 atom % D for NMR spectroscopy
Acetic acid-[D4]
Formula: D₃CCOOD
MW: 64.02 g/mol
Boiling Pt: 115.5
Melting Pt: 16
Flash Pt: 39
MDL Number: MFCD00051051
CAS Number: 1186-52-3
UN: 2789

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR