Dimethyl sulfoxide-[D₆] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

464071000
TS464071000EA 303 USD
TS464071000
Dimethyl sulfoxide-[D₆] 99.5 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MDL Number: MFCD00002090
CAS Number: 2206-27-1
UN: 1993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR